Learn More
CD4 Mouse anti-Rabbit, Unlabeled, Clone: KEN-4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 553625
Description
The KEN-4 antibody reacts with an antigen identified as CD4 on the majority of thymocytes and a subset of peripheral T lymphocytes. mAb KEN-4 immunoprecipitates, from thymocyte lysate, a major band of 50 kDa and a 42-kDa minor band under both reducing and non-reducing conditions. It also blocks the proliferative response of mesenteric lymph node cells to allogeneic splenocytes.

Specifications
CD4 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Japanese White rabbit thymocytes | |
0.5 mg | |
Immunology | |
Rabbit | |
IgG2a κ |
Flow Cytometry | |
KEN-4 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.