Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD4; Clone: L200; BD
Supplier: BD Biosciences 562842
Description
Host Species: Mouse
Clone L200
Isotype IgG1, κ
Species Reactivity Hu, Rh, Cyn, Bb
Flow Cytometry

Specifications
CD4 | |
L200 | |
Mouse | |
50 Tests | |
Primary | |
IgG1 |
Monoclonal | |
Brilliant Violet 421 | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction