Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD4, Clone: OX-38,

Catalog No. CBL1506MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL1506MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL1506MI Supplier MilliporeSigma™ Supplier No. CBL1506

Specifications

Antigen CD4
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OX-38
Conjugate Unconjugated
Formulation The monoclonal is presented as 0.5mg/1 mL in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_000616.3
Gene Symbols CD4, CD4mut
Host Species Mouse
Quantity 500 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.