Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, BD IMag-DM, Clone: L200, BD

Catalog No. BDB557767 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 109 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557767 1 x 109 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557767 Supplier BD Biosciences Supplier No. 557767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BD IMag™ anti-human CD4 Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD4-bearing T lymphocytes using the BD IMagnet™. The L200 antibody reacts with CD4 on human, rhesus and cynomolgus macaque, and baboon peripheral blood leukocytes; we have confirmed that the BD IMag™ particles can effectively separate the CD4-bearing cells of rhesus macaque blood. The distribution of CD4 on peripheral leukocytes is similar for both human and monkey. It is on the MHC class II-restricted T helper cells, with the majority of CD4-positive lymphocytes being CD8-negative. It is also found on most thymocytes and at low density on monocytes; it is not found on B or NK cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone L200
Conjugate BD IMag-DM
Description L3T4 ; T-cell surface antigen T4/Leu-3; W3/25 ; CD4 antigen (p55)
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HPB-ALL Cell Line
Purification Method Affinity Purified
Quantity 1 x 109 Cells
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.