Learn More
CD4 Mouse anti-Human, PerCP, Clone: L200, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550631
Description
The L200 monoclonal antibody specifically binds to the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. The L200 antibody also crossreacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both Rhesus and Cynomolgus Macaque monkeys. Crossreactivity on both lymphocytes and monocytes (weak) from Baboons is also observed. The distribution on lymphocytes is similar for both human and monkey cells, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.

Specifications
CD4 | |
Monoclonal | |
PerCP | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
L200 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HPB-ALL Cell Line | |
50 Tests | |
Immunology | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.