Learn More
CD4 Mouse anti-Human, PerCP, Clone: SK3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 347324
Description
The CD4 antibody, clone SK3, is derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human peripheral blood T lymphocytes.
The CD4 antibody recognizes a 55-kilodalton (kDa) glycoprotein that is present on T-helper/inducer lymphocytes and monocytes.

Specifications
CD4 | |
Monoclonal | |
3.2μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Human Peripheral Blood T Cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
SK3 | |
PerCP | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.