Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Rat, PE, Clone: OX-38, BD

Catalog No. BDB551397 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551397 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551397 Supplier BD Biosciences Supplier No. 551397

Mouse Monoclonal Antibody

The OX-38 antibody has been reported to react with the CD4 antigen on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), monocytes, macrophages, and some dendritic cells. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. The OX-38 antibody has been reported to bind to the same epitope of CD4 as that recognized by W3/25 mAb, which is a different epitope than that recognized by OX-35 mAb (Cat. No. 554837). In vivo blocking of some cell-mediated immune responses by mAb OX-38 has been reported. Injection of OX-38 mAb induces allograft unresponsiveness in rats, with varying results depending on the rat strain used (high or low responder). Furthermore, in vivo depletion of CD4+ lymphocytes has been reported with this antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone OX-38
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat thymocyte glycoproteins
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.