Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rat anti-Mouse, violetFluor™ 450, Clone: RM4-5, MilliporeSigma™

Catalog No. MABF575MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF575MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF575MI Supplier MilliporeSigma Supplier No. MABF575
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal antibody Anti-CD4 (mouse), violetFluor 450, clone RM4-5 validated for use in immunoprecipitation, immunohistochemistry, Immunofluorescence and Flow Cytometry for the detection of CD4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone RM4-5
Concentration 0.2mg/mL
Conjugate Violet Fluor 450 (V450)
Formulation Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols Cd4
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 25 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.