Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Mouse anti-Human, APC-H7, Clone: 5C3, BD

Catalog No. BDB561211 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561211 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561211 Supplier BD Biosciences Supplier No. 561211
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This 5C3 monoclonal antibody specifically binds to CD40, a 45-48 kDa type I integral membrane glycoprotein. CD40 is expressed on B lymphocytes, but is not expressed on terminally differentiated B cells. CD40 is also expressed by endothelial cells, basal epithelial cells and some epithelial cell carcinomas, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, and CD34+ hematopoietic progenitor cells. This antibody is useful for studying the roles played by CD40 in B-cell growth, proliferation, and differentiation including immunoglobulin isotype switching. Anti-CD40 antibodies have been reported to stimulate B-cell proliferation when costimulated with anti-µ, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry
Classification Monoclonal
Clone 5C3
Conjugate APC-H7
Description TNFRSF5; TNF receptor superfamily member 5; CD40L receptor; Bp50; p50
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.