Learn More
Description
The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumour necrosis factor/nerve growth factor superfamily and shows a significant homology to the Hodgkin's disease-associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.
Host Species: Mouse
Clone: 11E9
Isotype: IgG2b
Cross Reactivity: Human. Others not-tested.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding the entire external domain of human CD40.
Molecular Weight: 44kDa
Positive Control: Hodgkin's lymphoma
Cellular Localization: Cell membrane
Immunohistochemistry (Formalin/Paraffin)
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | CD40 Ab-1 |
| Classification | Monoclonal |
| Concentration | n/a |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding the entire external domain of human CD40. |
| Quantity | 7 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG2b |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.