Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD40, Clone: 11E9, Epredia™ LabVision

Catalog No. MS1090S Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
5 Slides
500 μL
7 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1090S 1 mL
MS1090S0 100 μL
MS1090S1 500 μL
MS1090R7 7 mL
MS1090PCS 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS1090S Supplier Epredia Supplier No. MS1090S

Mouse Monoclonal Antibody

The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumour necrosis factor/nerve growth factor superfamily and shows a significant homology to the Hodgkin's disease-associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.

Host Species: Mouse
Clone: 11E9
Isotype: IgG2b
Cross Reactivity: Human. Others not-tested.
Epitope: Extracellular domain
Immunogen: Recombinant protein encoding the entire external domain of human CD40.
Molecular Weight: 44kDa
Positive Control: Hodgkin's lymphoma
Cellular Localization: Cell membrane

Immunohistochemistry (Formalin/Paraffin)

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen CD40 Ab-1
Classification Monoclonal
Concentration n/a
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein encoding the entire external domain of human CD40.
Purification Method Hodgkin's lymphoma
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Content And Storage 2–8 degrees
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.