Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Mouse anti-Human, Unlabeled, Clone: 41, BD

Catalog No. BDB611363 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611363 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611363 Supplier BD Biosciences Supplier No. 611363
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD40, a member of the TNF receptor family, contains a cysteine-rich N-terminal domain and a Ser/Thr-rich region preceding the transmembrane domain. On B cells, signaling through CD40 induces cell growth and differentiation, mediates cell survival within the germinal center, and upregulates the expression of costimulatory and adhesion molecules, such as B7.1, B7.2, and ICAM-1. The interaction of CD40 on B cells and CD40L on activated CD4+ T cells is essential for immune functions, such as immuoglobulin class switching. Signal transduction through CD40 pathways involves interaction with proteins such as TRAFs (TRAF2, TRAF3, TRAF5, and TRAF6); Jak 3; and Tyr phosphorylation of proteins, such as Lyn, Syk, PI-3-kinase, STAT3, and STAT5. In TRAF2-deficient mice, CD40-mediated B cell proliferation and NFκB activation are defective. Ku70 and Ku80 associate with the membrane-proximal region of CD40 in human primary B cells and the engagement of CD40 leads translocation of Ku proteins to the nucleus. Thus, CD40 interacts with a variety of signal transducers which mediate its role in B cell survival, growth, differentiation, and immunoglobulin class switching.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Western Blot
Classification Monoclonal
Clone 41
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD40 aa. 71-187
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.