Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Mouse anti-Human, Brilliant Violet 711, Clone: 5C3, BD

Catalog No. BDB563397 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563397 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563397 Supplier BD Biosciences Supplier No. 563397
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This 5C3 monoclonal antibody specifically binds to CD40, a 45-48 kDa type I integral membrane glycoprotein. CD40 is expressed on B lymphocytes, but is not expressed on terminally differentiated B cells. CD40 is also expressed by endothelial cells, basal epithelial cells and some epithelial cell carcinomas, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, and CD34+ hematopoietic progenitor cells. This antibody is useful for studying the roles played by CD40 in B-cell growth, proliferation, and differentiation including immunoglobulin isotype switching. Anti-CD40 antibodies have been reported to stimulate B-cell proliferation when costimulated with anti-µ, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry
Classification Monoclonal
Clone 5C3
Conjugate Brilliant Violet 711
Description TNFRSF5; TNF receptor superfamily member 5; CD40L receptor; Bp50; p50
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.