Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD40 Clone: EA-5,

Catalog No. 217590100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
21-759-0100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-759-0100UG Supplier MilliporeSigma™ Supplier No. 217590100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD40 Clone: EA-5 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation

Human CD40 is a member of the tumor necrosis factor (TNF) family of cell surface receptors, and is found on all B cells (except plasma cells) and some epithelial, carcinoma, and lymphoid dendritic cells. Through binding of its ligand (CD40L/CD154/gp39), CD40 plays an important role in B-cell activation and isotype switching.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone EA-5
Conjugate Unconjugated
Formulation In 150mM NaCl, 100mM KCl, 50mM sodium phosphate, 0.5mg/mL gentamicin sulfate, pH 7.5.
Host Species Mouse
Immunogen transfected murine L cells expressing human CD40
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽44kDa CD40 protein in Raji cells.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.