Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Hamster anti-Mouse, FITC, Clone: HM40-3, BD

Catalog No. BDB553723 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553723 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553723 Supplier BD Biosciences Supplier No. 553723
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The HM40-3 antibody reacts with CD40, a 40-50-kDa glycoprotein expressed on B lymphocytes and other antigen-presenting cells. The CD40 molecule has a central role in B-cell growth and differentiation. Furthermore, interactions of CD40 with its ligand, CD154, are involved in the initiation and effector stages of cell-mediated immune responses. CD40 may be involved in the triggering of NK cells and NK-T cells. Soluble HM40-3 antibody stimulates splenic and peritoneal B cells to proliferate in vitro. This antibody also induces spleen B cells to express the costimulatory molecules CD80 (B7-1) and CD86 (B7-2). HM40-3 mAb has been demonstrated to inhibit the binding of soluble CD154 (gp39, CD40 Ligand) to soluble CD40 and to cell-surface CD40. This hamster mAb to a mouse leukocyte antigen has been observed to cross-react with similar populations of Lewis, Sprague-Dawley, and LOU16 rat leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry
Classification Monoclonal
Clone HM40-3
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen (BALB/c x NZB)F1 mouse-derived lymphoma WEHI-231
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.