Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD40L Clone: 24-31,

Catalog No. 217595100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
21-759-5100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-759-5100UG Supplier MilliporeSigma™ Supplier No. 217595100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects CD40L Clone: 24-31 in Human samples, and it is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization, Immuno Blotting

Human CD40 ligand (CD40L) is a member of the tumor necrosis factor (TNF) family expressed on the surface of activated T-cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40L
Applications Flow Cytometry, Immunoblot, Immunoblot, Immunohistochemistry (Frozen), Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 24-31
Conjugate Unconjugated
Formulation In 150mM NaCl, 100mM KCl, 50mM sodium phosphate, pH 7.5.
Host Species Mouse
Immunogen human sgp39 fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽36kDa CD40L (gp39) protein on activated CD3+ human PBL.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.