Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41b Mouse anti-Human, Unlabeled, Clone: HIP2, BD

Catalog No. BDB555468 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555468 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555468 Supplier BD Biosciences Supplier No. 555468
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIP2 monoclonal antibody specifically binds to CD41b. CD41b/CD41 is encoded by the ITGA2B gene and is also known as Platelet glycoprotein GPIIb and Integrin alpha IIb. CD41 is transmembrane glycoprotein that is post-translationally processed into a disulfide linked α-chain and β-chain. CD41 combines with CD61, otherwise known as Glycoprotein IIIa or β3 integrin, to form the CD41/CD61 (GPIIb/IIIa) complex. This complex is expressed on platelets and megakaryocytes and is involved in platelet activation, aggregation and binding to extracellular matrix proteins. Upon platelet activation, the CD41/CD61 complex can function as a receptor for fribrinogen, fibronectin and von Willebrand factor. In contrast to other CD41b antibodies, HIP2 does not detect anti-ZWa bound to GPIIb/IIIa. This antibody appears to be a weak aggregation inducer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD41b
Applications Flow Cytometry
Classification Monoclonal
Clone HIP2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description ITGA2B; CD41; GP2B; GPIIb; GPalpha IIb; Platelet glycoprotein GPIIb
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG3 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.