Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42a Mouse anti-Human, PerCP, Clone: BEB1, BD

Catalog No. BDB340537 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340537 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340537 Supplier BD Biosciences Supplier No. 340537
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD42a antibody, clone Beb1, is derived from hybridization of Sp2/0 mouse myeloma cells with spleen cells from BALB/c mice immunized with normal human platelets.

The CD42a antibody recognizes a 17 to 22-kilodalton (kDa) single-chain, platelet membrane glycoprotein, also known as gpIX that forms a noncovalent complex with gpIb and gpV. Glycoprotein Ib consists of an α–chain disulfide-linked to a β chain. Glycoproteins Ibα (140 to 145 kDa), Ibβ (24 to 25 kDa), and V (82 to 86 kDa) have been designated as CD42b, CD42c and CD42d respectively. CD42a has been reported to react with both gpIX and the gpIb-IX-V complex.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications Flow Cytometry
Classification Monoclonal
Clone BEB1
Concentration 12.5μg/mL
Conjugate PerCP
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human platelets
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.