Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43 Mouse anti-Human, Brilliant Violet 605, Clone: 1G10, BD

Catalog No. BDB563378 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563378 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563378 Supplier BD Biosciences Supplier No. 563378
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1G10 monoclonal antibody specifically binds to CD43, a 95-135 kDa sialoglycoprotein that is highly expressed on most human leucocytes. CD43 is encoded by the SPN gene and is also known as Sialophorin, Galactoglycoprotein (GALGP), Leukosialin, and Leukocyte sialoglycoprotein. The CD43 antigen is expressed on T cells, pre-B cells and activated B cells, NK cells and granulocytes, but is not present on resting peripheral blood B cells, red blood cells, platelets and non-hematopoietic cells. CD43 is enzymatically shed from leucocyte surfaces following activation by various stimuli. CD43 appears to be involved in intercellular interactions that regulate T, B, and NK cell functions. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections with TUF pretreatment.

For optimal and reproducible results, BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant dyes are used in the same experiment. Fluorescent dye interactions may cause staining artifacts which may affect data interpretation. The BD Horizon Brilliant Stain Buffer was designed to minimize these interactions. More information can be found in the Technical Data Sheet of the BD Horizon Brilliant Stain Buffer (Cat. No. 563794).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD43
Applications Flow Cytometry
Classification Monoclonal
Clone 1G10
Conjugate Brilliant Violet 605
Description SPN; Sialophorin; Galactoglycoprotein/GALGP; GPL115; LEUK; Leukosialin; LSN
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.