Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD43 Mouse anti-Human, Unlabeled, Clone: 1G10, BD

Catalog No. BDB555474 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555474 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555474 Supplier BD Biosciences Supplier No. 555474
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 1G10 monoclonal antibody specifically binds to CD43, a 95-135 kDa sialoglycoprotein that is highly expressed on most human leucocytes. CD43 is encoded by the SPN gene and is also known as Sialophorin, Galactoglycoprotein (GALGP), Leukosialin, and Leukocyte sialoglycoprotein. The CD43 antigen is expressed on T cells, pre-B cells and activated B cells, NK cells and granulocytes, but is not present on resting peripheral blood B cells, red blood cells, platelets and non-hematopoietic cells. CD43 is enzymatically shed from leucocytesurfaces following activation by various stimuli. CD43 appears to be involved in intercellular interactions that regulate T, B, and NK cell functions. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections with TUF pretreatment.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD43
Applications Flow Cytometry
Classification Monoclonal
Clone 1G10
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.