Learn More
CD43 Mouse anti-Human, NA/LE, Unlabeled, Clone: L60, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551457
Description
Anti-leukosialin monoclonal antibody (clone L60) recognizes a sialic acid-dependent determinant on leukosialin, a strongly anionic, highly o-linked glycosylated cell surface protein with a 45nm extracellular domain. Leukosialin, also know as sialophorin or CD43, is a major component of T lymphocyte, thymocyte, and neutrophil (PMN) membranes, and is also expressed on many different cells including monocytes, platelets, bone marrow stem cells, and plasma cells.

Specifications
CD43 | |
Monoclonal | |
0.5mg/mL | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
L60 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.