Learn More
Description
Host Species: Rat
Clone S7
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Mouse Plasmacytoma MOPC-315
Flow Cytometry
Specifications
Specifications
| Antigen | CD43 |
| Classification | Monoclonal |
| Clone | S7 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-Cyanine7 |
| Host Species | Rat |
| Immunogen | Mouse Plasmacytoma MOPC-315 |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.