Learn More
CD44 Mouse anti-Human, APC, Clone: G44-26 (also known as C26), BD
Mouse Monoclonal Antibody
$182.80 - $354.00
Specifications
Antigen | CD44 |
---|---|
Clone | G44-26 (also known as C26) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560890
|
BD Biosciences
560890 |
25 Tests |
Each for $182.80
|
|
|||||
BDB559942
|
BD Biosciences
559942 |
100 Tests |
Each for $354.00
|
|
|||||
Description
The G44-26 monoclonal antibody specifically binds to the 80-95 kDa, glycosylated type I transmembrane protein, also known as phagocytic glycoprotein-1 (Pgp-1). CD44 is the receptor for hyaluronic acid. CD44 is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is also called extracellular matrix receptor type III and has functional roles in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using citrate buffer pretreatment. This antibody recognizes the epitope 1 of CD44 antigen according to the HLDA workshop studies.
Specifications
CD44 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
G44-26 (also known as C26) | |
Monoclonal | |
Mouse | |
Human | |
IgG2b κ | |
Affinity Purified | |
Phagocytic glycoprotein 1; Pgp-1; H-CAM; Hermes; ECMR III; HUTCH-1 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.