Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD44 Rat anti-Human, Mouse, APC-Cy7, Clone: IM7, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABF581
Description
This rat monoclonal antibody Anti-CD44 (human/mouse), APC-Cy7, clone IM7 validated for use in western blotting, immunoprecipitation, immunohistochemistry and Flow Cytometry for the detection of CD44.
Specifications
CD44 | |
Monoclonal | |
0.2mg/mL | |
Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide. | |
Rat | |
Size Exclusion Chromatography | |
Primary | |
2-8°C, protect from prolonged exposure to light. Do not Freeze. | |
IgG2b κ |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
IM7 | |
APC-Cyanine7 | |
Cd44, Ly-24 | |
Proprietary immunogen | |
25 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction