Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD44, BV711; Clone: IM7; BD

Catalog No. BDB563971 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563971 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563971 Supplier BD Biosciences Supplier No. 563971
Add to Cart
Add to Cart

Host Species: Rat
Clone: IM7
Isotype: IgG1, κ
Species Reactivity: Ms
Immunogen: Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Classification Monoclonal
Clone IM7
Concentration 0.2mg/mL
Host Species Rat
Immunogen Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.