Learn More
Description
Host Species: Rat
Clone: IM7
Isotype: IgG1, κ
Species Reactivity: Ms
Immunogen: Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1
Flow cytometry
Specifications
Specifications
| Antigen | CD44 |
| Classification | Monoclonal |
| Clone | IM7 |
| Concentration | 0.2mg/mL |
| Host Species | Rat |
| Immunogen | Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1 |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.