Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Mouse anti-Human, NA/LE, Unlabeled, Clone: 515, BD

Catalog No. BDB550990 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550990 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550990 Supplier BD Biosciences Supplier No. 550990
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with CD44, and 85 kDa glycoprotein that is expressed on leukocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is a member of the hyaladherin family of hyaluronan-binding proteins, with structural similarities to selectins. It has a functional role in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. 515 monoclonal antibody has been reportedto be able to block cell adhesion to the hyaluronic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone 515
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Pgp-1, H-CAM, Ly24
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.