Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Rat anti-Mouse, Unlabeled, Clone: KM114, BD

Catalog No. BDB558739 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558739 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558739 Supplier BD Biosciences Supplier No. 558739
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The KM114 antibody reacts with an epitope on both alloantigens and all isoforms of the CD44 glycoprotein (Pgp-1, Ly-24). The KM114 hybridoma was produced at the same time as the published clone KM201, and the mAb recognizes a different epitope from that recognized by mAb IM7. The standard form of CD44, lacking variable exons and referred to as CD44H or CD44S, is widely expressed on hematopoietic and non-hematopoietic cells. CD44 isoforms encoded by variable exons are expressed on epithelial cells, but only at low levels on most leukocytes. Mice with the Ly-24.1 alloantigen (e.g., BALB/c, CBA/J, DBA/1, DBA/2) have relatively large subsets of CD44H+ T lymphocytes, while Ly-24.2 strains (e.g., A, AKR, CBA/N, C3H/He, C57BL, C57BR, C57L, C58, NZB, SJL, SWR, 129) have lower expression of CD44H on T cells. CD44 is a cell adhesion receptor, and its ligand, hyaluronate, is a common component of extracellular matrices. Differential glycosylation of CD44 influences its binding to hyaluronate. Additional ligands include the cell-surface form of CD74 and the cytokine osteopontin (Eta-1).10 Bone marrow- and thymus-derived progenitor cells capable of repopulating the thymus express CD44. In the periphery, the level of CD44 expression increases upon activation of B lymphocytes, CD4+ T cells, and CD8+ T cells; and memory cells can be recognized by their CD44[hi] phenotype. KM114 antibody can be used in ELISA to detect soluble CD44, and it is effective for in vitro blocking of hyaluronate recognition by CD44. It has been reported that KM114 mAb cross-reacts with CD44 of the Chinese hamster, Cricetulus griseus, but not human CD44.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone KM114
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Pgp-1, H-CAM, Ly-24
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen (C57BL/6 x DBA/2)F1 mouse bone marrow-derived stromal cell clone BMS2
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.