Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44 Mouse anti-Human, Unlabeled, Clone: L178, BD

Catalog No. BDB559046 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559046 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559046 Supplier BD Biosciences Supplier No. 559046
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with homing-associated cell adhesion molecule (HCAM), CD44, an integral membrane glycoprotein that exists in a variety of isoforms (CD44 variants). CD44 is synthesized as a 37 kDa polypeptide that is converted to a 80-90 kDa form by glycosylation via N- and O-linkages, or to a 180-200 kDa form by the addition of chondroitin sulfate. It is expressed on leukocytes, erythrocytes, epithelial cells and weakly on platelets. Depending on the cell type, CD44 may participate in a variety of functions including cell adhesion, motility and cell activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone L178
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.