Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD44 FITC, Clone: IM7,

Catalog No. MABF1556MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1556MI 25 μg
MABF1558MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1556MI Supplier MilliporeSigma™ Supplier No. MABF1556
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects CD44 Clone: IM7 in Human, Mouse samples, and it is validated for Flow Cytometry

The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or 'standard' form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Applications Flow Cytometry
Classification Monoclonal
Clone IM7
Conjugate FITC
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P15379
Gene Symbols Cd44, Ly-24
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.