Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD44 (H-CAM) Rat anti-Mouse, Phycoerythrin, Clone: KM201, MilliporeSigma™

Catalog No. FCMAB426PE Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB426PE 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB426PE Supplier MilliporeSigma Supplier No. FCMAB426PE

Rat Monoclonal Antibody

This Milli-Mark™ Anti-mouse CD44 (H-CAM)-PE Antibody, clone KM201 is validated for use in FC for the detection of mouse CD44 (H-CAM).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44 (H-CAM)
Applications Flow Cytometry
Classification Monoclonal
Clone KM201
Conjugate PE
Formulation Purified rat monoclonal IgG1 conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_001034240
Gene Symbols CD44, CD44R, CDW44, CDw44, CSPG8, ECMR-III, Epican, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, MUTCH-I, PGP-1, PGP-I, Pgp1
Host Species Rat
Immunogen Mouse bone marrow-derived stromal cells
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.