Learn More
CD44H Mouse anti-Rat, FITC, Clone: OX-49, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550974
Description
The OX-49 antibody reacts with the glycoprotein CD44H (also known as CD44s) expressed on most leukocytes, except for a subset of B lymphocytes, and at greatly increased levels on T- and B-cell blasts. The epitope recognized by OX-49 antibody has been mapped to a region on both the standard, CD44s, and the splice variant, CD44v, isoforms of CD44. However, recent reports indicate that OX-49 antibody cannot detect the CD44v isoform, possibly due to conformational changes in the epitope. CD44 is a cell adhesion receptor, and its ligand, hyaluronate, is a common component of extracellular matrices.

Specifications
CD44H | |
Monoclonal | |
0.5mg/mL | |
Pgp-1, H-CAM, CD44s | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
OX-49 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat T blasts from mixed lymphocyte reactions | |
0.1 mg | |
Immunology | |
Rat | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.