Learn More
MilliporeSigma™ anti-CD44v6, exon v6 ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB2080/DEL
Description
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
CD44v6, exon v6 | |
Unconjugated | |
Synthetic peptide of human CD44v6 | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
100 μg | |
Cell Structure | |
Bovine, Canine, Human, Mouse, Pig, Rabbit |