Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD44v7 Rabbit anti-Bovine, Canine, Human, Mouse, Pig, Rat, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. AB2083MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2083MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2083MI Supplier MilliporeSigma Supplier No. AB2083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect CD44v7 using this Anti-CD44v7 Antibody, exon v7 validated for use in ELISA, flow cytometry, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44v7
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 10mM sodium phosphate, pH 7.5 with 150mM NaCl with 0.01% sodium azide.
Gene Symbols CD44; LHR; HUTCH-I; CDW44; CDw44; ECMR-III; Epican; MGC10468; CD44R; MDU2; PGP-1; CSPG8; Pgp1; IN; MUTCH-I; MDU3; MIC4; MC56; HCELL
Host Species Rabbit
Immunogen Synthetic peptide of human CD44v7
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000610.3; NM_001001389.1; NM_001001390.1; NM_001001391.1; NM_001001392.1
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.