Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD45.2, Alexa Fluor 700, Clone: 104, R&D Systems™

Catalog No. FAB8448N025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8448N025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8448N025 Supplier R&D Systems Supplier No. FAB8448N025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Antigen CD45.2
Applications Flow Cytometry
Classification Monoclonal
Clone 104
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias B220, CD 45, CD45, cd45 antigen, CD45.2, CD45R, GP 180, GP180, Human homolog of severe combined immunodeficiency due to PTPRC deficiency, L CA, LCA, Leukocyte common antigen, LY 5, Ly5, Ly5.2, Ly-5.2, Protein tyrosine phosphatase receptor type C, Protein tyrosine phosphatase receptor type c polypeptide, PTPRC, RT7.2, SCID due to PTPRC deficiency, T200, T200 glycoprotein, T200 leukocyte common antigen
Gene Symbols PTPRC
Host Species Mouse
Immunogen B10.S mouse thymocytes and splenocytes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19264
Test Specificity Detects mouse cells expressing the CD45.2 allotype in flow cytometry. Clone 104 does not detect the mouse CD45.1 alloantigen.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.