Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Human, AlexaFluor™488, Clone: HI30, MilliporeSigma™

Catalog No. MABF323MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF323MI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF323MI Supplier MilliporeSigma Supplier No. MABF323

Mouse Monoclonal Antibody

Anti-CD45 (human) Antibody, AlexaFluor™488, clone HI30 is a highly specific mouse monoclonal antibody, that targets CD45 and has been tested in Flow Cytometry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone HI30
Conjugate Alexa Fluor 488
Formulation Purified mouse conjugate antibody in buffer containing PBS (pH 7.2) with up to 0.1% sodium azide and 0.2% (w/v) BSA.
Gene Symbols PTPRC, CD45
Host Species Mouse
Immunogen Proprietary immunogen
Purification Method Affinity
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.