Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse, Unlabeled, Clone: 69, BD

Catalog No. BDB610266 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610266 150 μg
BDB610265 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610266 Supplier BD Biosciences Supplier No. 610266
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45 (also known as leukocyte common antigen, GP180, or T200) is found on all hematopoietic cells except those of red cell origin. The N-terminal domain is a large glycosylated extracellular region of variable length (390-542 amino acids) derived by the alternative splicing of at least three exons (4, 5, or 6). This variation accounts for the different isoforms of CD45 (180-220 kDa). The remainder of the molecule includes a short transmembrane region followed by a large highly conserved cytoplasmic domain of 702 residues. The CD45 cytoplasmic domain possesses an intrinsic protein tyrosine phosphatase (PTPase) activity. This region contains two PTPase domains (domains I and II) which share 40% homology. Both domains are required for optimal PTPase activity and show significant homology with other receptor-like and cytoplasmic PTPases. CD45 plays a critical role in antigen receptor-induced responses of T and B cells. In B cells, CD45 can exist as part of a larger multisubunit complex composed of several tyrosine phosphorylated proteins, including the Src-family PTK, Lyn, or it can exist as the membrane immunoglobulin-associated MB-1/B29 heterodimer. In T cells, CD45 associates with and dephosphorylates both lck and fyn. In addition, CD45 can associate with membrane proteins involved in T cell activation, such as CD2, Thy-1, TCR, and CD26.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Western Blot
Classification Monoclonal
Clone 69
Concentration 250μg/mL
Conjugate Unconjugated
Description Leukocyte Common Antigen; GP180; T200
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.