Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse, PE-CF594, Clone: DO58-1283, BD

Catalog No. BDB562394 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562394 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562394 Supplier BD Biosciences Supplier No. 562394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

D058-1283 is a CD45 monoclonal antibody specific for non-human primate leucocytes, developed using Rhesus peripheral whole blood as immunogen. It does not cross react with human leucocytes. This antibody reacts with Baboon, Rhesus and Cynomolgus Macaque leucocytes in a similar pattern as seen with CD45 binding to the Leukocyte Common Antigen on human cells. Immunophenotypic analysis shows that D058-1283 binds to lymphocytes, monocytes and granulocytes of non-human primate blood samples. This antibody is able to block the binding of monoclonal antibody TÜ116; a reported anti-human CD45 antibody that cross-reacts with nonhuman primate leucocytes. In Western blot analysis, the D058-1283 antibody identifies a band of approximately 180-200 kDa molecular weight .

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone DO58-1283
Conjugate PE-CF594
Description Pan Leukocyte, NHP-specific
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rhesus peripheral whole blood
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.