Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Human, Mouse, Rat, Clone: F10-89-4, MilliporeSigma™

Catalog No. 051410MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-141-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-141-0MI Supplier MilliporeSigma Supplier No. 051410
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD45 Antibody, clone F10-89-4 detects level of CD45 and has been published and validated for use in flow cytometry, western blot, immunofluoresence, immunoprecipitation, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F10-89-4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a presented in phosphate buffered saline containing 10mM sodium azide and 1mg/mL bovine serum albumin.
We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_002838.3
Gene Symbols PTPRC, L-CA, LCA, CD45, LY5, T200, Glycoprotein, B220, GP180, EC 3.1.3.48
Host Species Mouse
Immunogen Purified T cells from human lymph nodes
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 24 months. Avoid repeated freeze/thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.