Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD45; Clone: HI30; BD
$221.00 - $438.00
Specifications
Antigen | CD45 |
---|---|
Clone | HI30 |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Host Species | Mouse |
Description
Host Species: Mouse
Clone: HI30
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Peripheral Blood Leucocytes
Flow cytometry
Specifications
CD45 | |
Monoclonal | |
Mouse | |
Human | |
Primary |
HI30 | |
Brilliant Ultraviolet 395 | |
RUO | |
Human Peripheral Blood Leucocytes | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title