Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Rat anti-Mouse, Clone: IBL-5/25, MilliporeSigma™

Catalog No. 051416MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-141-6MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-141-6MI Supplier MilliporeSigma Supplier No. 051416
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-CD45 Antibody, clone IBL-5/25 is an antibody against CD45 for use in flow cytometry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone IBL-5/25
Conjugate Unconjugated
Formulation Purified rabbit monoclonal IgG in phosphate buffered saline containing 10mM sodium azide.
Gene Accession No. NP_002829.2
Gene Symbols CD45-AP , LPAP, MGC138602, MGC138603
Host Species Rat
Immunogen Mouse lymph node cells.
Purification Method Protein G purfied
Quantity 500 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.