Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 (Leukocyte Common Antigen) Mouse anti-Human, APC, Clone: 2D1, BD

Catalog No. BDB340943 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340943 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340943 Supplier BD Biosciences Supplier No. 340943
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45 clone 2D1 is derived from the hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human peripheral blood mononuclear cells (PBMCs).

The CD45 antibody (anti–HLe-1) recognizes members of the T200 family of human leucocyte antigens with molecular mass of 180 to 220 kilodaltons (kDa).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone 2D1
Concentration 25μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.