Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Human, PE-CF594, Clone: HI30, BD

Catalog No. BDB562312 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562312 25 Tests
BDB562279 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB562312 Supplier BD Biosciences Supplier No. 562312
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone HI30
Conjugate PE-CF594
Description PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Peripheral Blood Leucocytes
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.