Learn More
CD45R/B220 Rat anti-Mouse, Biotin, Clone: RA3-6B2, BD
Rat Monoclonal Antibody
$96.40 - $327.50
Specifications
Antigen | CD45R |
---|---|
Clone | RA3-6B2 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The RA3-6B2 antibody reacts with an epitope on the extracellular domain of CD45 glycoprotein which is dependent upon the expression of exon A and specific carbohydrate residues. It is expressed on B lyphocytes at all stages from pro-B through mature and activated B cell, but it is decreased on plasma cells and a subset of memory B cells. Levels of expression of CD45R/B220 on the B-cell lineage appear to be developmentally regulated. It is also found on the abnormal T cells involved in the lymphadenopathy of lpr/lpr and gld/gld mutant mice, on lytically active subsets of lymphokine-activated killer cells (NK cells and non-MHC-restricted CTL), on apoptotic T lymphocytes of mice injected with bacterial superantigen, on a population of NK-cell precursors in the bone marrow, and on B-lymphocyte, T-lymphocyte, and macrophage progenitors in fetal liver. The CD45R/B220 antigen is not on hematopoietic stem cells, naive T lymphocytes, or MHC-restricted CTL. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. CD45R/B220 is commonly used as a pan B-cell marker; however, CD19 expression, detected by mAb 1D3, is reported to be more restricted to the B-cell lineage. mAb RA3-6B2 has been reported to enhance isotype switching during in vitro B-cell responses and to inhibit in vivo B-cell responses. Cross-reaction of RA3-6B2 mAb with activated human T lymphocytes has been observed. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: RA3-6B2
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse Pre-B Tumor
Cell Separation, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
CD45R | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
RA3-6B2 | |
Flow Cytometry | |
Biotin | |
RUO | |
Human, Mouse | |
Mouse Pre-B Tumor | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.