Learn More
Description
Host Species: Rat
Clone: RA3-6B2
Isotype: IgG2a, κ
Species Reactivity: Ms
Immunogen: Mouse Abelson Leukemia Virus-Induced pre-B tumor cells
Flow cytometry
Specifications
Specifications
| Antigen | CD45R |
| Classification | Monoclonal |
| Clone | RA3-6B2 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Ultraviolet 395 |
| Host Species | Rat |
| Immunogen | Mouse Abelson Leukemia Virus-Induced pre-B tumor cells |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.