Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD45R (B220) Rat anti-Mouse, FITC, Clone: RA3-3A1/6.1, MilliporeSigma™

Catalog No. FCMAB228FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB228FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB228FMI Supplier MilliporeSigma Supplier No. FCMAB228F

Rat Monoclonal Antibody

Milli-Mark™ Anti-mouse CD45R (B220)-FITC Antibody, clone RA3-3A1/6.1 is an antibody against mouse CD45R (B220) for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45R (B220)
Applications Flow Cytometry
Classification Monoclonal
Clone RA3-3A1/6.1
Conjugate FITC
Formulation Purified rat monoclonal IgM conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_001104786
Gene Symbols Ptprc, Ly-5
Host Species Rat
Immunogen Animals were immunized with the mouse cell line RAW 112. Spleen cells were fused with the S194/5.XX0.BU.1 myeloma.
Purification Method Ion Exchange
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.