Learn More
CD45R (B220) Rat anti-Human, Mouse, PE-Cy5, Clone: RA3-6B2, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | CD45R (B220) |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | RA3-6B2 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-Cyanine5 |
| Formulation | Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide. |
| Gene Symbols | Ptprc, Ly-5 |
| Host Species | Rat |
| Immunogen | Proprietary immunogen |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.