Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Rat anti-Mouse, Biotin, Clone: 14.8, BD

Catalog No. BDB557460 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557460 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557460 Supplier BD Biosciences Supplier No. 557460

Rat Monoclonal Antibody

The 14.8 clone has been reported to react with an exon A-dependent epitope of the CD45 protein, which is found at high density on B cells and at low density on peripheral T cytotoxic/suppressor cells and a very small subset of thymocytes. Nearly all B-lineage cells, including B-cell precursors in fetal liver and adult bone marrow and Ig-secreting cells, but not hematopoietic stem cells or myeloid progenitors, have been reported to be detectable by mAb 14.8. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus, differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. mAb 14.8 has been reported to enhance the proliferative effect of PHA on purified spleen T cells, possibly by replacing a signal normally delivered by accessory cells, to enhance isotype switching during in vitro B-cell responses, and to inhibit antigen-induced p21 [ras] activation.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry
Classification Monoclonal
Clone 14.8
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Radiation-induced NZC mouse B lymphoma WEHI-279
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.