Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RA Mouse anti-Human, Mouse, Rat, Clone: MEM 56, MilliporeSigma™

Catalog No. 051413MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-141-3MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-141-3MI Supplier MilliporeSigma Supplier No. 051413
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD45RA Antibody, Clone MEM 56 detects level of CD45RA and has been published and validated for use in flow cytometry, immunofluoresence, immunohistochemistry (paraffin), immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RA
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM 56
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in phosphate buffered saline containing 10mM sodium azide and 1mg/mL bovine serum albumin.
We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_002838.3
Gene Symbols PTPRC, L-CA, LCA, CD45, LY5, T200, Glycoprotein, B220, GP180, EC 3.1.3.48
Host Species Mouse
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.