Learn More
CD45RA Mouse anti-Human, V450, Clone: 5H9, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561220
Description
The 5H9 monoclonal antibody specifically binds to the human form of the leukocyte common antigen (LCA), CD45RA. This clone also crossreacts with CD45RA expressed by peripheral blood lymphocytes, monocytes and some granulocytes from rhesus macaque, baboon, and cynomolgus monkeys. The observed frequency of CD45RA positive lymphocytes is higher in the non-human primate compared to what is seen with normal human donor lymphocytes.
The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Specifications
CD45RA | |
Monoclonal | |
Violet Fluor 450 (V450) | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5H9 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.